AuftragsverfolgungKontakt
Microplates, 96-well
Microplates, 96-well
96-Well-Platte, flacher Boden, steril, Klar, 370 µl, 96
Catalog # KART2620/S
Supplier:   KARTELL
Microplates, 96-well
Catalog # KART2620/S
Supplier:  KARTELL
Microplates, 96-well
Microplates, 96-well
96-Well-Platte, flacher Boden, steril, Klar, 370 µl, 96
Price
SFr. 108.00
Log in for availability
1 * 50 ST
Product Details & Documents
Multiple-well plates (96 sample wells) in clear PS. Available in three shapes: flat bottom, U bottom and V bottom wells.
  • Aseptic or sterile
  • Single-wrapped
Ordering information: PS covers are sold separately.
Specifications
  • Anz. Wells:
    96
  • Volumen:
    370 µl
  • Farbe:
    Klar
  • Bezeichnung:
    96-Well-Platte, flacher Boden, steril
Accessories
Product Information
  • Catalog #
    Bezeichnung
    Availability
    Price
  • KART2623
    Deckel für 96-Well-Mikrotiterplatten, PS, aseptisch
    Log in for availability
    1 * 50 ST - SFr. 83.30
Product Information
  • Catalog #
    Bezeichnung
    Availability
    Price
  • KART2623/S
    Deckel für 96-Well-Mikrotiterplatten, PS, steril
    Log in for availability
    1 * 50 ST - SFr. 87.00