AuftragsverfolgungKontakt
J.T.Baker® DILUT-IT™, Potassium Phosphate, Dissolution Media Concentrate
Dissolution media concentrate, DILUT-IT, potassium phosphate, pH 7,5, 6×1,9 l, Poly bottle, 1,9 L
  D016-04
 :   Avantor
J.T.Baker® DILUT-IT™, Potassium Phosphate, Dissolution Media Concentrate
  D016-04
 :  Avantor
 :  
J.T.Baker® DILUT-IT™, Potassium Phosphate, Dissolution Media Concentrate
J.T.Baker® DILUT-IT™, Potassium Phosphate, Dissolution Media Concentrate
Dissolution media concentrate, DILUT-IT, potassium phosphate, pH 7,5, 6×1,9 l, Poly bottle, 1,9 L
 :  
SFr. 93.30
1 * 1,9 L
DILUT-IT is a ready-to-use dissolution media concentrate, which is pre-measured and only need to be diluted with purified water.
DILUT-IT™ dissolution media concentrates are produced from raw materials in accordance with USP/EP/BP/JP guidelines under validated processes in Avantor Performance Materials' ISO certified manufacturing facilities
  • Packung:
    Poly bottle
  • VE:
    1,9 L
  • Bezeichnung:
    Dissolution media concentrate, DILUT-IT, potassium phosphate, pH 7,5, 6×1,9 l