J.T.Baker® DILUT-IT™, Potassium Phosphate, Dissolution Media Concentrate
Dissolution media concentrate, DILUT-IT, potassium phosphate, pH 7,5, 6×1,9 l, Poly bottle, 1,9 L
: Avantor
DILUT-IT is a ready-to-use dissolution media concentrate, which is pre-measured and only need to be diluted with purified water.
DILUT-IT™ dissolution media concentrates are produced from raw materials in accordance with USP/EP/BP/JP guidelines under validated processes in Avantor Performance Materials' ISO certified manufacturing facilities
- Packung:Poly bottle
- VE:1,9 L
- Bezeichnung:Dissolution media concentrate, DILUT-IT, potassium phosphate, pH 7,5, 6×1,9 l