Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
 

PIG® Poly utility trays

Poly utility trays, 33 L, 650×965×64 mm
  NEWPPAK661
 :   NEW PIG
PIG® Poly utility trays
  NEWPPAK661
 :  NEW PIG
PIG® Poly utility trays
PIG® Poly utility trays

Poly utility trays, 33 L, 650×965×64 mm

 
SFr. 104.00
   
1 * 1 ST
 

Flat-bottom poly trays are the economical way to store small parts. Great for lining your benches and shelving..

  • Light-coloured, flat-bottom design makes it easy to see screws, springs and other small parts
  • All-poly construction resists most chemicals; will not rust or corrode
  • Rotationally molded spill tray is ideal for applications that would compromise injected or thermoformed trays
  • Made from FDA-compliant resins for containment of food and cosmetic products

Square modular design allows efficient use of space on bench or shelves for process parts.

 
  • Kapazität:
    33 L
  • Bezeichnung:
    Poly utility trays
  • Seq:
    04
  • B×T×H:
    650×965×64 mm