Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Office desks

Office desks

Desk top, NS16, walnut, 1600×800×720 mm
  132-0753
Office desks
  132-0753
Office desks
Office desks

Desk top, NS16, walnut, 1600×800×720 mm

 
 
   
1 * 1 ST
 

The table top is 25 mm thick, made of walnut, melamine coated according to DIN 68765. The C-shaped frame is made of round tubes with a diameter of 35 or 45 mm. The main advantage of this line desks is that they can be assembled according to the user's need.

  • Ergonomic and convenient
  • Solid construction
  • Adjustable height, using levellers on legs: ± 5 mm

Delivery information: Furniture is delivered flat-packed. The enclosed instructions make assembly easy, even for the layman. Assembly service available for an extra charge.

 
  • Bezeichnung:
    Desk top, NS16, walnut
  • B×T×H:
    1600×800×720 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.