Membrane filters, Millipore Express® PLUS
HPWP, 0,45 µm, 25 mm
Katalognr. MILFHPWP02500
Anbieter: Merck
Direkt zu
Product Details & Documents
These PES white membrane filters provides ultrafast filtration of tissue culture media, additives, buffers and other aqueous solutions. This high throughput, low protein-binding membrane is also used in many of our ready-to-use sterile filtration devices.
- Ultra-fast filtration and high throughput shorten process time
- Can be sterilised by autoclave, ethylene oxide, or gamma irradiation
Widely used for sterile filtration, buffer filtration and tissue culture media filtration.
Surface: plain
Bacterial endotoxins: 0,5 EU/ml
Gravimetric extractables: <0,5%
Technische Daten
- Ø:25 mm
- Porengröße:0,45 µm
- Typ:HPWP
- Seq:07
- Cat. No.:MILFHPWP02500
- Luftdurchsatz:40 ml/min cm²
- Stärke:140 µm
- Wasserdurchfluss:23 ml/min/cm² (690 mbar)
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



