Membrane filters, Millipore Express® PLUS
HPWP, 0,45 µm, 13 mm
Katalognr. MILFHPWP01300
Anbieter: Merck
Direkt zu
Product Details & Documents
These PES white membrane filters provides ultrafast filtration of tissue culture media, additives, buffers and other aqueous solutions. This high throughput, low protein-binding membrane is also used in many of our ready-to-use sterile filtration devices.
- Ultra-fast filtration and high throughput shorten process time
- Can be sterilised by autoclave, ethylene oxide, or gamma irradiation
Widely used for sterile filtration, buffer filtration and tissue culture media filtration.
Surface: plain
Bacterial endotoxins: 0,5 EU/ml
Gravimetric extractables: <0,5%
Technische Daten
- Ø:13 mm
- Porengröße:0,45 µm
- Typ:HPWP
- Seq:06
- Cat. No.:MILFHPWP01300
- Luftdurchsatz:40 ml/min cm²
- Stärke:140 µm
- Wasserdurchfluss:23 ml/min/cm² (690 mbar)



