Ribbons for IP™ label printing system
IP-R-6002, Farbband-Serie 6000, Harz-Ausführung, Breite 83 mm, 300 m, Schwarz
BRDY066226
: Brady
These IP® printer ribbons are high density ribbon with excellent solvent resistance, good heat resistance and super scratch resistance and have ribbon spindle.
- Farbe:Schwarz
- Bezeichnung:IP-R-6002, Farbband-Serie 6000, Harz-Ausführung, Breite 83 mm
- Länge:300 m
- Seq:07
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.