Ethylenglycol ≥99%
Glasflasche für Feststoffe, 2.500 g
Anbieter: Thermo Fisher Scientific
Direkt zu
Product Details & Documents
Purity: 99% CAS No.: 107-21-1 Molecular Formula: C2H6O2 Formula Weight: 62.07 Flash Point: 119°(246°F) Refractive Index: 1.4310 Odour: Sweet Melting Point: -13° Boiling Point: 196-198° Beilstein Reference: 505945 Merck Reference: 14,3798 MDL No.: MFCD00002885
Synonyme: Ethane-1,2-diol, 1,2-Dihydroxy ethane, 1,2-Ethanediol, Glycol
- CAS Nummer:107-21-1
- Formel:HOCH₂CH₂OH
- MDL-Nummer:MFCD00002885
- Molekulargewicht:62.07 g/mol
- Merck-Index:14,3798
- Siedepunkt:196…198 °C
- Schmelzender Platingehalt:–13 °C
- Dichte:1,113
- Blitz Pt:119 °C (246 °F)
- Lagertemperatur:Raumtemperatur
- EINECS:203-473-3
- ERREICHEN:01-2119456816-28
Documents
Zertifikate
Suche nach ChargennummerALFAA11591.30_TDS_EN
Product specification
Organic Building Blocks and Intermediates
Application Notes
Technische Daten
Technische Daten
- Packung:Glasflasche für Feststoffe
- VE:2.500 g
- Cat. No.:A11591.0E
- Umweltzertifizierung:Chem21 Solvent Selection Guide recommended by default and after discussion
- Umweltfreundlichkeit:Unbedenklicher für Mensch und Umwelt
Technische Daten
Consigli
I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".
Altrimenti riceverai suggerimenti generici..



