Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
 

Supelco® COD CRM, standard solutions

CSB-Standardlösung, 200 mg/L, ± 4 mg/L , 1.14895; 1.14690; 1.14541, Plastic bottle, 3 - 5, 100 mL
Anbieter:   Merck
Supelco® COD CRM, standard solutions
Katalognr. 1.25030.0100
Anbieter:  Merck
CAS Nummer:  
Supelco® COD CRM, standard solutions
Supelco® COD CRM, standard solutions

CSB-Standardlösung, 200 mg/L, ± 4 mg/L , 1.14895; 1.14690; 1.14541, Plastic bottle, 3 - 5, 100 mL

CAS Nummer:  
Preis
46.10 SFr.
Einloggen für Verfügbarkeit
1 * 100 mL
Product Details & Documents
  • Diluted standard solutions that are ready-to-use for accurate measurements
  • Traceable to SRM from NIST and USP
  • Full certificates of analysis with comprehensive quality documentation
  • Information on concentration and expanded measurement uncertainty
  • Save time and minimize errors

Method used for Uncertainty Measurement (Accuracy in the table below)= Photometry

 : Traceable to SRM from NIST and USP.

Technische Daten

Technische Daten

  • Konzentration:
    200 mg/L
  • Packung:
    Plastic bottle
  • VE:
    100 mL
  • Bezeichnung:
    CSB-Standardlösung
  • Genauigkeit:
    ± 4 mg/L
  • pH-Bereich:
    3 - 5
  • Für Test-Kit(s):
    1.14895; 1.14690; 1.14541
  • Cat. No.:
    1.25030.0100
  • M-Clarity Qualitätsstufe:
    MQ200

Technische Daten