Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Supelco® COD CRM, standard solutions

Supelco® COD CRM, standard solutions

CSB-Standardlösung, 20,0 mg/L, ± 0,7 mg/L , 1.14560; 1.01796, Plastic bottle, 4 - 6, 100 mL
Codice catalogo 1.25028.0100
Nome del fornitore:   Merck
Supelco® COD CRM, standard solutions
Codice catalogo 1.25028.0100
Nome del fornitore:  Merck
Numero CAS:  
Supelco® COD CRM, standard solutions
Supelco® COD CRM, standard solutions

CSB-Standardlösung, 20,0 mg/L, ± 0,7 mg/L , 1.14560; 1.01796, Plastic bottle, 4 - 6, 100 mL

Numero CAS:  
Price
SFr. 49.10
Log in per disponibilità
1 * 100 mL
Product Details & Documents
  • Diluted standard solutions that are ready-to-use for accurate measurements
  • Traceable to SRM from NIST and USP
  • Full certificates of analysis with comprehensive quality documentation
  • Information on concentration and expanded measurement uncertainty
  • Save time and minimize errors

Method used for Uncertainty Measurement (Accuracy in the table below)= Photometry

Certifications: Traceable to SRM from NIST and USP.

Specifiche

Specifiche

  • Konzentration:
    20,0 mg/L
  • Packung:
    Plastic bottle
  • VE:
    100 mL
  • Bezeichnung:
    CSB-Standardlösung
  • Genauigkeit:
    ± 0,7 mg/L
  • pH-Bereich:
    4 - 6
  • Für Test-Kit(s):
    1.14560; 1.01796
  • Cat. No.:
    1.25028.0100
  • M-Clarity Qualitätsstufe:
    MQ200

Specifiche

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.