Stoppers, for test tubes
Grün, 11 - 12 mm
Katalognr. RATI351490G
Anbieter: RATIOLAB
Direkt zu
Product Details & Documents
PE.
- Very tight seal due to two sealing rings
- Available in natural and four more colors
Technische Daten
- Ø unten:11 - 12 mm
- Farbe:Grün
- Seq:03