AuftragsverfolgungKontakt
Biotin reagents, OneQuant™ HOOK™ | Avantor
Biotin reagents, OneQuant™ HOOK™
OneQuant™ HOOK™ Sulfo-NHS-SS-biotin, 1 mg, 1 KIT
Katalognr. 786-699
Anbieter:   G-Biosciences
Biotin reagents, OneQuant™ HOOK™
Katalognr. 786-699
Anbieter:  G-Biosciences
CAS Nummer:  
Biotin reagents, OneQuant™ HOOK™
Biotin reagents, OneQuant™ HOOK™
OneQuant™ HOOK™ Sulfo-NHS-SS-biotin, 1 mg, 1 KIT
CAS Nummer:  
Preis
SFr. 247.00
Einloggen für Verfügbarkeit
1 * 1 KIT
Product Details & Documents
The OneQuant™ format prevents loss of reagent due to repeated weighing as each vial contains only 1 - 2 mg HOOK™ Biotin Reagent.
Technische Daten
Technische Daten
  • VE:
    1 KIT
  • Bezeichnung:
    OneQuant™ HOOK™ Sulfo-NHS-SS-biotin, 1 mg