Stopper, HDPE, KIMBLE®
13 mm
Katalognr. SCERSP28160R-13
Anbieter: DWK Life Sciences
Direkt zu
Product Details & Documents
HDPE, linear, ideal for flasks that are grounded to [ST] stopper dimensions.
- Yellow and blue stopper with a closed bottom
- Enlarged flange is designed to protect the neck if the glassware is tipped over
Technische Daten
- Ø unten:13 mm
- Seq:02