AuftragsverfolgungKontakt
Screw caps | Avantor
Screw caps
GL 18
Katalognr. VITL102597
Anbieter:   VITLAB
Screw caps
Katalognr. VITL102597
Anbieter:  VITLAB
Screw caps
Screw caps
GL 18
Preis
SFr. 10.40
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
PFA, transparent
  • Especially suitable for use in trace analysis
  • Ideal for sensitive and valuable samples
  • High thermal stability, from –200 °C to +260 °C, autoclavable
  • Chemical inertness against nearly all chemicals
  • Easy to clean
For sealing all PFA containers with GL threads or buttress threads.
Technische Daten
  • Gewinde:
    GL 18