Spoon spatulas
210 mm
Katalognr. JUCHVL04014
Anbieter: JUCHHEIM Laborgerate GmbH
Direkt zu
Product Details & Documents
Double ended spoon spatulas made from stainless steel.
- One broad spatula end, one spoon end
Technische Daten
- Länge:210 mm
- Seq:03