Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
 

Retort rings

Ring with bosshead, 266 g, 130 mm
  JUCHVL04970
Retort rings
  JUCHVL04970
Retort rings

Ring with bosshead, 266 g, 130 mm

 
 
   
1 * 1 ST
 

Steel, epoxy coated.

  • Available with or without bosshead
  • Bosshead is made of malleable iron
 
  • Ext.Ø:
    130 mm
  • Bezeichnung:
    Ring with bosshead, 266 g
  • Seq:
    10
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.