Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Retort rings

Retort rings

Ring with bosshead, 210 g, 70 mm
Katalognr. JUCHVL04950
Retort rings
Katalognr. JUCHVL04950
Retort rings

Ring with bosshead, 210 g, 70 mm

Preis
SFr. 25.70
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents

Steel, epoxy coated.

  • Available with or without bosshead
  • Bosshead is made of malleable iron
Technische Daten
  • Ext.Ø:
    70 mm
  • Bezeichnung:
    Ring with bosshead, 210 g
  • Seq:
    08
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.