Retort rings
Ring with bosshead, 165 g, 50 mm
Katalognr. JUCHVL04940
Anbieter: JUCHHEIM Laborgerate GmbH
Direkt zu
Product Details & Documents
Steel, epoxy coated.
- Available with or without bosshead
- Bosshead is made of malleable iron
Technische Daten
- Ext.Ø:50 mm
- Bezeichnung:Ring with bosshead, 165 g
- Seq:07



