Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Retort rings

Retort rings

Ring without bosshead, 346 g, 180 mm
Katalognr. JUCHVL04932
Retort rings
Katalognr. JUCHVL04932
Retort rings

Ring without bosshead, 346 g, 180 mm

Preis
Preis anfragen
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents

Steel, epoxy coated.

  • Available with or without bosshead
  • Bosshead is made of malleable iron
Technische Daten
  • Ext.Ø:
    180 mm
  • Bezeichnung:
    Ring without bosshead, 346 g
  • Seq:
    06
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.