Retort rings
Ring without bosshead, 230 g, 130 mm
Katalognr. JUCHVL04920
Anbieter: JUCHHEIM Laborgerate GmbH
Direkt zu
Product Details & Documents
Steel, epoxy coated.
- Available with or without bosshead
- Bosshead is made of malleable iron
Technische Daten
- Ext.Ø:130 mm
- Bezeichnung:Ring without bosshead, 230 g
- Seq:04
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



