Hook bossheads
Hook bosshead with M8 stainless steel thumb bolt, 13 mm, Edelstahl
Katalognr. JUCHVL0486018/8
Anbieter: JUCHHEIM Laborgerate GmbH
Direkt zu
Product Details & Documents
Hook bossheads made of steel, epoxy coated or stainless steel with brass pressure block.
Technische Daten
- Spannweite:13 mm
- Bezeichnung:Hook bosshead with M8 stainless steel thumb bolt
- Material:Edelstahl
- Seq:02
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.