Double bossheads, cross shape
Double bosshead, cross shape, with M8 stainless steel thumb bolt, Eisen, 16 mm
Katalognr. JUCHVL04850S
Anbieter: JUCHHEIM Laborgerate GmbH
Direkt zu
Product Details & Documents
Bossheads made from either epoxy coated malleable iron or from stainless steel.
Technische Daten
- Bezeichnung:Double bosshead, cross shape, with M8 stainless steel thumb bolt
- Material:Eisen
- Für Stab Ø:16 mm