Spray bottles
500 ml, 206 mm
Katalognr. MURRP014/22P513/14
Anbieter: MURRAY S
Direkt zu
Product Details & Documents
HDPE, translucent.
Designed to allow comfortable grip and minimise fatigue. Adjustable from fine spray to single jet. Delivery 1,4 ml per stroke.
Technische Daten
- Kapazität:500 ml
- Höhe:206 mm