Microplates, 96-Well
White, poly-D-lysine, lid with condensation rings, 25 - 340 µl, -, 392 µl
Katalognr. GREI655944BE
Direkt zu
Artikel gestrichen
Das Produkt wurde eingestellt. Wir haben jedoch ähnliche Alternativprodukte gefunden.
Alternativen
Product Details & Documents
PS, without lid. A large number of different 96-well microplates are available in a wide variety of surface treatments with a choice of well profile, and with either solid or µClear® bottom. All plates are free of DNase, RNase and human DNA, and are non-pyrogenic.
Technische Daten
- Empf. Arbeitsvolumen:25 - 340 µl
- Steril:-
- Bezeichnung:White, poly-D-lysine, lid with condensation rings
- Seq:16
- Gesamtvolumen:392 µl