Chlorphenolrot
Glass bottle for solids, 5 g
Chlorphenolrot
Katalognr. ABCRAB132629.0005
CAS Nummer: 4430-20-0
Direkt zu
Artikel gestrichen
Das Produkt wurde eingestellt. Wir haben jedoch ähnliche Alternativprodukte gefunden.
Alternativen
Chlorophenol Red 1 * 5 g Katalognr.B21623.06
Product B21623.06 is a replacement for ABCRAB132629.0005 and has the same packaging and specifications, but may be a different quantity.
Product Details & Documents
CAS No.: 4430-20-0 Molecular Formula: C19H12Cl2O5S Formula Weight: 432.28 Beilstein Reference: 354053 MDL No.: MFCD00005877
Synonyme: 3,3-Bis(3-chloro-4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide, 4,4'-(3H-2,1-Benzoxathiol-3-ylidene)bis[2-chlorophenol] S,S-dioxide
- CAS Nummer:4430-20-0
- Formel:C₁₉H₁₂Cl₂O₅S
- MDL-Nummer:MFCD00005877
- Molekulargewicht:423.27 g/mol
- Schmelzender Platingehalt:∼261 °C
- Lagertemperatur:Raumtemperatur
- EINECS:224-619-2
Dokumente
Technische Daten
Technische Daten
- Packung:Glass bottle for solids
- VE:5 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



