Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
 

Natriumpyruvat ≥99%

Glasflasche für Feststoffe, 50 g
Catalog # ABCRAB126319.0050
Natriumpyruvat ≥99%
Catalog # ABCRAB126319.0050
Supplier:  
CAS Number:  113-24-6
Natriumpyruvat ≥99%
Natriumpyruvat ≥99%

Glasflasche für Feststoffe, 50 g

CAS Number:  113-24-6
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
Sodium pyruvate, 99% 1 * 50 g Catalog #A11148.18

 

Product Details & Documents

Purity: 99% CAS No.: 113-24-6 Molecular Formula: C3H3NaO3 Formula Weight: 110.04 Melting Point: 300° Beilstein Reference: 3568341

Synonyms:  2-Ketopropionic acid sodium salt, 2-Oxopropionic acid sodium salt, Sodium 2-oxopropanoate, Pyruvic acid sodium salt, α-Ketopropionic acid sodium salt
  • CAS Number:113-24-6
  • Formula:CH₃COCOONa
  • MDL Number:MFCD00002586
  • Molecular Weight:110.04 g/mol
  • Melting Pt:>300 °C
  • Density:1,718 g/cm³ (20 °C)
  • Storage Temperature:Raumtemperatur
  • EINECS:204-024-4
Specifications

Specifications

  • Packung:
    Glasflasche für Feststoffe
  • VE:
    50 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.