Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
4-Brombenzonitril ≥98%

4-Brombenzonitril ≥98%

10 g
Catalog # ABCRAB110356.0010
Toxic
Irritant
4-Brombenzonitril ≥98%
Catalog # ABCRAB110356.0010
Supplier:  
CAS Number:  623-00-7
4-Brombenzonitril ≥98%
4-Brombenzonitril ≥98%

10 g

CAS Number:  623-00-7
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
4-Bromobenzonitrile, 98% 1 * 10 g Catalog #A10914.09

Product A10914.09 is a replacement for ABCRAB110356.0010 and has the same packaging and specifications, but may be a different quantity.

Product Details & Documents

Purity: 98+% CAS No.: 623-00-7 Molecular Formula: C7H4BrN Formula Weight: 182.03 Flash Point: 130°(266°F) Melting Point: 111-114° Boiling Point: 235-237° Beilstein Reference: 2041518 UN No.: 3439 MDL No.: MFCD00001811

Synonyms:  p-Bromobenzonitrile
  • CAS Number:623-00-7
  • Formula:C₇H₄BrN
  • MDL Number:MFCD00001811
  • Molecular Weight:182.02 g/mol
  • Boiling Pt:235…237 °C
  • Melting Pt:111…114 °C
  • Density:1,562
  • Flash Pt:130 °C (266 °F)
  • Storage Temperature:Raumtemperatur
  • EINECS:210-764-9
  • UN:3439
  • ADR:6.1,III
Specifications

Specifications

  • VE:
    10 g

Specifications