Stirrer guides, universal, PTFE
Stirrer guide, with standard tapper 19/22 NS, 6 mm, 96 mm
Katalognr. COWI005.101.1.7
Anbieter: COWIE
Direkt zu
Product Details & Documents
Stirrer guide features composite PTFE/PEEK seal and a glass ball-bearing for rigidity and smoothness of operation.
- Resistant to chemicals
- Anti-whip and reduced vibration
They have an NPT screw thread fitting for direct assembly into PTFE reactor lids or NPT extension adapters and standard taper cone fittings are for use in standard glass joints.
Technische Daten
- Bezeichnung:Stirrer guide, with standard tapper 19/22 NS
- Höhe:96 mm
- Seq:08
- Schaft - Ø:6 mm
Consigli
I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".
Altrimenti riceverai suggerimenti generici..



