Magnetic stirring bars, PTFE, triangular, Spinstar®
Triangular, Reacti-Vial™ small reaction vials, 0,3 and 1,0 ml
Katalognr. HYPETS-16010
Anbieter: Thermo Fisher Scientific
Magnetic stirring bars, PTFE, triangular, Spinstar®
Katalognr. HYPETS-16010
Anbieter: Thermo Fisher Scientific
Direkt zu
Product Details & Documents
Triangular stir bars can be used for faster reaction times and smooth mixing of small samples in conical-bottom vials of the Thermo Scientific™ Reacti-Therm™ Derivatization System. The stir bars provide simultaneous stirring of samples in multiple vials when used with a Reacti-Therm Heating/Stirring Module equipped with appropriate Reacti-Block Aluminium Blocks.
Technische Daten
- Für:Reacti-Vial™ small reaction vials, 0,3 and 1,0 ml
- Typ:Triangular
- Seq:01