Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Magnetic stirring bars, PTFE, triangular, Spinstar®
Magnetic stirring bars, PTFE, triangular, Spinstar®
Triangular, Reacti-Vial™ small reaction vials, 3,0, 5,0 and 10 ml
Catalog # HYPETS-16000
Magnetic stirring bars, PTFE, triangular, Spinstar®
Catalog # HYPETS-16000
Magnetic stirring bars, PTFE, triangular, Spinstar®
Triangular, Reacti-Vial™ small reaction vials, 3,0, 5,0 and 10 ml
Price
SFr. 235.00
Log in for availability
1 * 6 ST
Product Details & Documents
Triangular stir bars can be used for faster reaction times and smooth mixing of small samples in conical-bottom vials of the Thermo Scientific™ Reacti-Therm™ Derivatization System. The stir bars provide simultaneous stirring of samples in multiple vials when used with a Reacti-Therm Heating/Stirring Module equipped with appropriate Reacti-Block Aluminium Blocks.
Specifications
  • Für:
    Reacti-Vial™ small reaction vials, 3,0, 5,0 and 10 ml
  • Typ:
    Triangular
  • Seq:
    02
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.