Extender tools
Extender tool, Acetalpolymer, Standard knurl
Katalognr. UPCHP-298
Anbieter: UPCHURCH SCIENTIFIC
Direkt zu
Product Details & Documents
These tools made from aluminium or Delrin® can be used to tighten most knurled nuts in hard to reach places. For precise tightening, the extender tools listed with ¼" hex drives are designed to adapt to any torque wrench with a female ¼" socket.
Technische Daten
- Bezeichnung:Extender tool
- Material:Acetalpolymer
- Seq:02
- Verwendung mit Kopfeinsätzen:Standard knurl