Extender tools
Extender tool, with 1/4" hex drive, Aluminium, Headless knurl
Codice catalogo UPCHP-292
Nome del fornitore: UPCHURCH SCIENTIFIC
Vai a
Product Details & Documents
These tools made from aluminium or Delrin® can be used to tighten most knurled nuts in hard to reach places. For precise tightening, the extender tools listed with ¼" hex drives are designed to adapt to any torque wrench with a female ¼" socket.
Specifiche
- Bezeichnung:Extender tool, with 1/4" hex drive
- Material:Aluminium
- Seq:06
- Verwendung mit Kopfeinsätzen:Headless knurl