Disposable cuvettes
Makro, 350 – 800 nm, 45 mm, 4 ml, 10 mm, PS
Catalog # SEMA2532
Supplier: SEMADENI
Jump to:
Product Details & Documents
Disposable cuvettes for photometric measurements in the visible range (350 to 800 nm).
Specifications
- Lichtweg:10 mm
- Typ:Makro
- Volumen:4 ml
- Fenstermaterial:PS
- Höhe:45 mm
- Seq:002
- Messbereich:350 – 800 nm
Accessories
Product Information
- Catalog #BezeichnungAvailabilityPrice
- SEMA2533LDPE-Kappen für Küvetten mit quadratischer ÖffnungLog in for availability1 * 100 ST - SFr. 21.80