AuftragsverfolgungKontakt
Storage bags | Avantor
Storage bags
Storage bags, transparent, with ground weld., 0,05 mm, 200×370 mm
Katalognr. SEMA2447
Anbieter:   SEMADENI
Storage bags
Katalognr. SEMA2447
Anbieter:  SEMADENI
Storage bags
Storage bags, transparent, with ground weld., 0,05 mm, 200×370 mm
Preis
20.90 SFr.
Einloggen für Verfügbarkeit
1 * 100 ST
Product Details & Documents
Storage bags in transparent LDPE, with bottom weld.
It has bottom weld. They are also suitable for liquids but without side weld.
Verpackung: Minimum quantity:100 pieces
Technische Daten
  • Bezeichnung:
    Storage bags, transparent, with ground weld.
  • Abmessungen:
    200×370 mm
  • Seq:
    05
  • Stärke:
    0,05 mm