Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Clamps

Clamps

Rod, aluminium, for attaching clamps, length 200 mm, Ø 12 m
Katalognr. SEMA1086
Anbieter:   SEMADENI
Clamps
Katalognr. SEMA1086
Anbieter:  SEMADENI
Clamps
Clamps

Rod, aluminium, for attaching clamps, length 200 mm, Ø 12 m

Preis
Preis anfragen
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents

Clamps made of polyacetal (POM), in light blue.

  • Chemical and temperature resistant
  • Sticks (80 mm high) can be cut to the desired height
Technische Daten
  • Bezeichnung:
    Rod, aluminium, for attaching clamps, length 200 mm, Ø 12 m
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.