HPLC syringes
Autosampler syringe, -, 250 µl, - mm, AS1000, AS3000
Katalognr. 555-8225
Anbieter: Thermo Fisher Scientific
Direkt zu
Product Details & Documents
These HPLC syringes are standard fixed needle syringes for Rheodyne/Valco™ injectors.
- Easy and accurate
- Reproducible manual injections
Applications include industrial chromatography.
Technische Daten
- Für:AS1000, AS3000
- Gauge:-
- Volumen:250 µl
- Bezeichnung:Autosampler syringe
- Länge:- mm
- Seq:07