Narrow pattern forceps
Edelstahl, 145 mm, Serrated, Gerade, 2,25×1,45 mm, 2,25 mm
Katalognr. FSCI11002-14
Marketsource
Anbieter: FINE SCIENCE TOOLS
Direkt zu
Product Details & Documents
Technische Daten
- Länge:145 mm
- Material:Edelstahl
- Seq:05
- Spitzentyp:Serrated
- Spitzengröße:2,25×1,45 mm
- Spitzenbreite:2,25 mm
- Spitzenform:Gerade