Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Hardened fine scissors | Avantor

Hardened fine scissors

Sharp/sharp, curved tips, 110 mm, 26 mm
Katalognr. FSCI14091-11
Marketsource
Anbieter:   FINE SCIENCE TOOLS
Hardened fine scissors
Katalognr. FSCI14091-11
Anbieter:  FINE SCIENCE TOOLS
Marketsource
Hardened fine scissors
Hardened fine scissors

Sharp/sharp, curved tips, 110 mm, 26 mm

Preis
SFr. 72.37
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents

These scissors are made of stainless steel.

Technische Daten
  • Typ:
    Sharp/sharp, curved tips
  • Länge:
    110 mm
  • Seq:
    07
  • Spitzenlänge:
    26 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.