S&T CAF-4 clamp applying forceps
Not lockable, Edelstahl, 140 mm
Katalognr. FSCI00072-14
Marketsource
Anbieter: FINE SCIENCE TOOLS
Direkt zu
Product Details & Documents
Technische Daten
- Typ:Not lockable
- Länge:140 mm
- Material:Edelstahl
- Seq:02