Biemer vessel spring scissors
Blunt, straight tips, 130 mm, 2 mm, 2 mm
FSCI15032-13
Marketsource
These stainless steel scissors are designed to securely cut vessels without slipping.
- Typ:Blunt, straight tips
- Länge:130 mm
- Seq:02
- Spitzengröße:2 mm
- Spitzenlänge:2 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



