Surgical scissors, blunt
Blunt/blunt, curved tips, 130 mm, 42 mm
Catalog # FSCI14003-13
Marketsource
Supplier: FINE SCIENCE TOOLS
Jump to:
Product Details & Documents
These scissors are made of stainless steel.
Specifications
- Typ:Blunt/blunt, curved tips
- Länge:130 mm
- Seq:03
- Spitzenlänge:42 mm