Surgical scissors, blunt
Blunt/blunt, straight tips, 165 mm, 53 mm
Catalog # FSCI14000-16
Marketsource
Supplier: FINE SCIENCE TOOLS
Jump to:
Product Details & Documents
These scissors are made of stainless steel.
Specifications
- Typ:Blunt/blunt, straight tips
- Länge:165 mm
- Seq:08
- Spitzenlänge:53 mm