Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Surgical scissors, blunt

Surgical scissors, blunt

Blunt/blunt, straight tips, 200 mm, 60 mm
Katalognr. FSCI14000-20
Marketsource
Anbieter:   FINE SCIENCE TOOLS
Surgical scissors, blunt
Katalognr. FSCI14000-20
Anbieter:  FINE SCIENCE TOOLS
Marketsource
Surgical scissors, blunt
Surgical scissors, blunt

Blunt/blunt, straight tips, 200 mm, 60 mm

Preis
48.55 SFr.
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents

These scissors are made of stainless steel.

Technische Daten
  • Typ:
    Blunt/blunt, straight tips
  • Länge:
    200 mm
  • Seq:
    12
  • Spitzenlänge:
    60 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.