Fine scissors, sharp-blunt
Sharp/blunt, curved tips, 100 mm, 27 mm
Katalognr. FSCI14029-10
Marketsource
Anbieter: FINE SCIENCE TOOLS
Direkt zu
Product Details & Documents
These scissors are made of stainless steel.
Technische Daten
- Typ:Sharp/blunt, curved tips
- Länge:100 mm
- Seq:01
- Spitzenlänge:27 mm