Chlorine analysis kit
PD 250 total chlorine set, includes dispenser and reagent vial, 1 SET
Katalognr. 705-0966
Anbieter: Lovibond
Direkt zu
Product Details & Documents
The PD250 is designed for easy and controlled dosage of DPD Powder reagents. One click gives the exact amount of reagent required for a 10 ml sample. The PD 250 is the perfect alternative to the Powder Packs for those carrying out a number of tests, saving time while also reducing the amount of packaging waste.
- Easy handling, Precise dosage
- 5 years reagent shelf life (unopened vial)
- Determination of chlorine according to ISO 7393-2:2000 (free + total), 250 tests
Lieferinformationen: PD 250 in carton, including one reagent vial and instruction manual.
Technische Daten
Technische Daten
- VE:1 SET
- Bezeichnung:PD 250 total chlorine set, includes dispenser and reagent vial
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



