Pipette tips, bulk, Corning® DeckWorks™
Product Details & Documents
These bulk pipette tips are compatible with DeckWorks racks and packaged in resealable polyethylene bags. Tip sizes include 10, 200 and 1000 μl and offer maximum savings.
- System is designed to allow you to maximise storage space and help reduce the production and waste of plastic materials
- Both the hinged racks and reload decks within this system are manufactured with recycled plastic
- Tips are made in a controlled molding environment to ensure accuracy and precision across all tip volumes
Medical grade resins and production excellence allow for full certification as: nonpyrogenic, human DNA-free, RNase-/DNase-free, sterility assuarance level 10⁻³. All tips are manufactured from polypropylene.
The entire DeckWorks system is comprised of 7 sizes of graduated pipette tips which are longer than standard tips to minimise the contamination issues often associated with traditional pipette tip styles. Sizes include 10, 20, 100, 200, 200, extended length, 300, and 1000 µl tips.
Ordering information: Corning DeckWorks pipette tips are also available in hinged rack, barrier hinged rack, reload, reload starter kit, and tip station configurations.
Documents
Certificates
Search by Lot NumberSpecifications
- Verpackt:Lose verpackt; 768 Spitzen/Packung
- Steril:-
- Volumen:100 - 1000 µl
- Spitzentyp:Runde Spitze
- Spitzenfarbe:Transparent
- Cat. No.:CORN4114
- Umweltfreundlichkeit:Abfallreduzierend