Electrolyte filling solution for Ag/AgCl reference electrodes, Supelco®
Potassium chloride solution, Reference electrodes, Glasflasche, 1 L
Anbieter: Merck
Electrolyte filling solution for Ag/AgCl reference electrodes, Supelco®
Katalognr. 60137-1L
Anbieter: Merck
CAS Nummer:
Direkt zu
Product Details & Documents
~3 M Potassium chloride solution saturated with silver chloride (~0,16 M AgCl).
Dokumente
Zertifikate
Suche nach ChargennummerTechnische Daten
Technische Daten
- Packung:Glasflasche
- VE:1 L
- Bezeichnung:Potassium chloride solution
- Für:Reference electrodes
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.