Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Maxima™ First strand cDNA synthesis kit
Maxima™ First strand cDNA synthesis kit
Maxima® First Strand cDNA Synthesis Kit for RT-qPCR, 200 reactions, 1 KIT
Catalog # K1642
Maxima™ First strand cDNA synthesis kit
Catalog # K1642
CAS Number:  
Maxima™ First strand cDNA synthesis kit
Maxima™ First strand cDNA synthesis kit
Maxima® First Strand cDNA Synthesis Kit for RT-qPCR, 200 reactions, 1 KIT
CAS Number:  
Price
SFr. 2’200.00
Log in for availability
1 * 1 KIT
Product Details & Documents
The Maxima™ First Strand cDNA Synthesis Kit is a convenient system optimised for use as a first step in two step RT-qPCR. The kit includes the Maxima™ Reverse Transcriptase, which is an advanced enzyme derived by in vitro evolution of M-MuLV RT for RT-qPCR application. The enzyme features high thermostability, robustness and increased synthesis rates in RT-qPCR.
  • Increased reaction temperatures – up to 60 °C
  • Premixed components for convenient use in RT-qPCR
  • Increased synthesis rate – complete cDNA synthesis in 15 minutes
  • High sensitivity and specificity
The Maxima™ First Strand cDNA Synthesis Kit is capable of cDNA synthesis at elevated temperatures (50 to 60 °C) and completion of the reverse transcription reaction in 15 minutes. In addition, the kit offers highly convenient composition, where Maxima™ Reverse Transcriptase and RiboLock™ RNase Inhibitor are premixed, while all other reaction components: reaction buffer, dNTPs and primers (random hexamers and Oligo(dT)₁₈) are premixed in another tube as a 5X reaction mix.
Delivery information: Kit includes Maxima™ enzyme mix, 5X reaction mix, nuclease-free water, and detailed protocol.
Specifications
Specifications
  • VE:
    1 KIT
  • Bezeichnung:
    Maxima® First Strand cDNA Synthesis Kit for RT-qPCR, 200 reactions
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.