Maxima™ First strand cDNA synthesis kit
Product Details & Documents
The Maxima™ First Strand cDNA Synthesis Kit is a convenient system optimised for use as a first step in two step RT-qPCR. The kit includes the Maxima™ Reverse Transcriptase, which is an advanced enzyme derived by in vitro evolution of M-MuLV RT for RT-qPCR application. The enzyme features high thermostability, robustness and increased synthesis rates in RT-qPCR.
- Increased reaction temperatures – up to 60 °C
- Premixed components for convenient use in RT-qPCR
- Increased synthesis rate – complete cDNA synthesis in 15 minutes
- High sensitivity and specificity
The Maxima™ First Strand cDNA Synthesis Kit is capable of cDNA synthesis at elevated temperatures (50 to 60 °C) and completion of the reverse transcription reaction in 15 minutes. In addition, the kit offers highly convenient composition, where Maxima™ Reverse Transcriptase and RiboLock™ RNase Inhibitor are premixed, while all other reaction components: reaction buffer, dNTPs and primers (random hexamers and Oligo(dT)₁₈) are premixed in another tube as a 5X reaction mix.
Lieferinformationen: Kit includes Maxima™ enzyme mix, 5X reaction mix, nuclease-free water, and detailed protocol.
Technische Daten
Technische Daten
- VE:1 KIT
- Bezeichnung:Maxima® First Strand cDNA Synthesis Kit for RT-qPCR, 50 reactions
Technische Daten
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen..