Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
 

Replacement needles for syringes RTC/RSH

Replacement needle for 25 - 500 µl syringes, 23, 50 mm, 0,63 mm, Side hole/dome
  SGEA038736
Replacement needles for syringes RTC/RSH
  SGEA038736
Replacement needles for syringes RTC/RSH

Replacement needle for 25 - 500 µl syringes, 23, 50 mm, 0,63 mm, Side hole/dome

 
SFr. 87.80
   
1 * 2 ST
 
  • Ext.Ø:
    0,63 mm
  • Gauge:
    23
  • Spitzentyp:
    Side hole/dome
  • Bezeichnung:
    Replacement needle for 25 - 500 µl syringes
  • Länge:
    50 mm
  • Seq:
    13
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.